Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286408.1 | NP_001273337.1 | 5 | 991 | ||
NM_001286409.1 | NP_001273338.1 | 2 | 696 | |||
NM_198081.4 | NP_932347.2 | 6 | 1206 | |||
XM_011535703.2 | XP_011534005.1 | 6 | 1104 | |||
XM_011535704.2 | XP_011534006.1 | 6 | 982 | |||
XM_011535706.1 | XP_011534008.1 | 5 | 948 | |||
XM_011535707.2 | XP_011534009.1 | 6 | 1225 | |||
XM_011535709.2 | XP_011534011.1 | 2 | 1138 | |||
XM_011535710.2 | XP_011534012.1 | 4 | 815 | |||
XM_017010677.1 | XP_016866166.1 | 6 | 1273 | |||
XM_017010678.1 | XP_016866167.1 | 6 | 1186 | |||
XM_017010679.1 | XP_016866168.1 | 6 | 1084 | |||
XM_017010680.1 | XP_016866169.1 | 6 | 1152 | |||
XM_017010681.1 | XP_016866170.1 | 6 | 1062 | |||
XM_017010682.1 | XP_016866171.1 | 6 | 1276 | |||
XM_017010683.1 | XP_016866172.1 | 5 | 945 | |||
XM_017010684.1 | XP_016866173.1 | 6 | 1278 | |||
XM_017010685.1 | XP_016866174.1 | 5 | 1072 | |||
XM_017010686.1 | XP_016866175.1 | 5 | 1057 | |||
GenBank mRNA | AK074125.1 | - | 4 | 2531 | ||
AK092812.1 | - | 2 | 403 | |||
AK093571.1 | - | 5 | 991 | |||
AK303562.1 | - | 6 | 1193 | |||
BC033286.1 | - | 6 | 973 | 4509724 | ||
BG767121.1 | - | 2 | 696 |