Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198279.3 | NP_938020.2 | 3 | 1209 | ||
XM_005274473.3 | XP_005274530.1 | 4 | 1766 | |||
XM_005274474.3 | XP_005274531.1 | 4 | 1766 | |||
XM_005274475.3 | XP_005274532.1 | 4 | 1766 | |||
XM_011545475.2 | XP_011543777.1 | 4 | 1376 | |||
XM_011545476.2 | XP_011543778.1 | 4 | 1343 | |||
XM_011545477.2 | XP_011543779.1 | 4 | 1764 | |||
XM_011545478.2 | XP_011543780.1 | 4 | 1766 | |||
XM_011545479.2 | XP_011543781.1 | 4 | 1765 | |||
XM_011545482.2 | XP_011543784.1 | 4 | 1764 | |||
XM_017029390.1 | XP_016884879.1 | 4 | 1764 | |||
XM_017029391.1 | XP_016884880.1 | 4 | 1766 | |||
XM_017029392.1 | XP_016884881.1 | 4 | 1764 | |||
GenBank mRNA | AK094661.1 | - | 1 | 34 | ||
AK127709.1 | - | 2 | 825 | |||
AL833278.1 | - | 2 | 1059 | |||
BC126172.1 | - | 1 | 344 | |||
BC143906.1 | - | 1 | 344 |