Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147156.3 | NP_671512.1 | 7 | 1373 | ||
XM_005269675.1 | XP_005269732.1 | 2 | 1065 | |||
XM_011539582.2 | XP_011537884.1 | 4 | 977 | |||
XM_011539583.2 | XP_011537885.1 | 2 | 854 | |||
XM_011539584.2 | XP_011537886.1 | 4 | 977 | |||
GenBank mRNA | AB154421.1 | - | 3 | 756 | ||
AK026683.1 | - | 3 | 836 | |||
AY280959.1 | - | 7 | 1339 | |||
AY312431.1 | - | 1 | 565 | |||
AY332650.1 | - | 7 | 1248 | |||
BC042899.1 | - | 1 | 634 | 4342258 | ||
FM210200.1 | - | 1 | 551 | |||
FN870365.1 | - | 1 | 551 | |||
FN908045.1 | - | 1 | 551 | |||
FN908146.1 | - | 2 | 767 | |||
KR020015.1 | - | 272 | ||||
KR020016.1 | - | 272 |