Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 6 | 2643 | ||
NM_018136.4 | NP_060606.3 | 6 | 2643 | |||
GenBank mRNA | AF509326.1 | - | 6 | 2386 | ||
AK001380.1 | - | 2 | 318 | |||
AK125107.1 | - | 6 | 2643 | |||
AK226178.1 | - | 6 | 2576 | |||
AY099891.1 | - | 4 | 1168 | |||
AY099893.1 | - | 4 | 1168 | |||
AY367065.1 | - | 6 | 2386 | |||
AY971956.1 | - | 6 | 2386 | |||
BC034607.1 | - | 2 | 262 |