Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF520800.1 | - | 7 | 565 | ||
AF520805.1 | - | 5 | 437 | |||
AF520806.1 | - | 5 | 410 | |||
AF548113.1 | - | 6 | 642 | |||
AF548115.1 | - | 2 | 136 | |||
AF548116.1 | - | 3 | 222 | |||
AF548121.1 | - | 132 | ||||
AY309094.1 | - | 8 | 646 | |||
BC113541.1 | - | 6 | 504 | 8322593 | ||
BC113543.1 | - | 6 | 504 | 8322595 | ||
BC143837.1 | - | 7 | 516 | 9052352 |
Gene Symbol: LOC107983965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011612.1 | XP_016867101.1 | 8 | 862 |