Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153274.2 | NP_695006.1 | 4 | 652 | ||
XM_017001020.1 | XP_016856509.1 | 4 | 1900 | |||
XM_017001021.1 | XP_016856510.1 | 6 | 879 | |||
XM_017001022.1 | XP_016856511.1 | 6 | 2003 | |||
XM_017001023.1 | XP_016856512.1 | 6 | 2003 | |||
GenBank mRNA | AF440757.1 | - | 4 | 603 | ||
AY515707.1 | - | 4 | 603 | |||
BC101823.1 | - | 4 | 652 | 8069329 |