Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153274.2 | NP_695006.1 | 2 | 272 | ||
XM_017001020.1 | XP_016856509.1 | 2 | 1520 | |||
XM_017001021.1 | XP_016856510.1 | 4 | 499 | |||
XM_017001022.1 | XP_016856511.1 | 4 | 1623 | |||
XM_017001023.1 | XP_016856512.1 | 4 | 1623 | |||
GenBank mRNA | AF440757.1 | - | 2 | 223 | ||
AY515707.1 | - | 2 | 223 | |||
BC101823.1 | - | 2 | 272 | 8069329 |