Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198868.1 | NP_001185797.1 | 18, 19 | 2058 | ||
NM_001198869.1 | NP_001185798.1 | 18, 19 | 2027 | |||
NM_005186.3 | NP_005177.2 | 18, 19 | 2043 | |||
NR_040008.1 | 17, 18 | 1973 | ||||
XM_006718698.1 | XP_006718761.1 | 18, 19 | 1958 | |||
XM_011545292.1 | XP_011543594.1 | 18, 19 | 1969 | |||
GenBank mRNA | AK097277.1 | - | 1, 2 | 1438 | ||
AK301538.1 | - | 17, 18 | 1865 | |||
AY550975.1 | - | 17, 18 | 1866 | |||
BC008751.2 | - | 18, 19 | 1994 | 3347480 | ||
BC017200.2 | - | 18, 19 | 1880 | 3629894 | ||
BC075862.1 | - | 18, 19 | 1961 | 5223130 | ||
X04366.1 | - | 18, 19 | 2004 |