Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EYA3 SIRNA Assays ›
Gene Symbol:
EYA3
Gene Name:
EYA transcriptional coactivator and phosphatase 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 27970344 - 28088671 on Build GRCh38
UniGene ID:
Hs.185774
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4910
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EYA3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282560.1
NP_001269489.1
12, 13
1195
NM_001282561.1
NP_001269490.1
12, 13
1195
NM_001282562.1
NP_001269491.1
12, 13
1253
NM_001990.3
NP_001981.2
13, 14
1333
NR_104214.1
14, 15
1401
XM_006710449.3
XP_006710512.1
14, 15
1369
XM_011540999.2
XP_011539301.1
14, 15
1372
XM_011541001.2
XP_011539303.1
13, 14
1330
XM_011541003.2
XP_011539305.1
13, 14
1232
XM_011541004.2
XP_011539306.1
13, 14
1229
XM_017000652.1
XP_016856141.1
14, 15
1373
XM_017000653.1
XP_016856142.1
15, 16
1493
XM_017000654.1
XP_016856143.1
13, 14
1330
XM_017000655.1
XP_016856144.1
14, 15
1486
XM_017000656.1
XP_016856145.1
13, 14
1233
XM_017000657.1
XP_016856146.1
12, 13
1225
XM_017000658.1
XP_016856147.1
12, 13
1250
XM_017000659.1
XP_016856148.1
11, 12
1109
GenBank mRNA
AK289805.1
-
13, 14
1292
AK295745.1
-
12, 13
1189
AK298129.1
-
12, 13
1250
AK303664.1
-
12, 13
1177
BC029500.1
-
14, 15
1361
5272319
BC041667.1
-
12, 13
1122
5743906
BX648945.1
-
13, 14
1302
U81602.1
-
8, 9
932
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107910, XM_006239025, XM_006239024, XM_006239027, XM_006239028, and others.
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
visual perception
anatomical structure morphogenesis
response to ionizing radiation
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
positive regulation of DNA repair
protein tyrosine phosphatase activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0