Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318812.1 | NP_001305741.1 | 3 | 460 | ||
NM_001318813.1 | NP_001305742.1 | 3 | 562 | |||
NM_001318814.1 | NP_001305743.1 | 4 | 644 | |||
NM_001318815.1 | NP_001305744.1 | 3 | 405 | |||
NM_001318816.1 | NP_001305745.1 | 3 | 730 | |||
NM_173810.3 | NP_776171.1 | 2 | 648 | |||
GenBank mRNA | AF289605.1 | - | 4 | 1013 | ||
AF451992.1 | - | 2 | 589 | |||
AF451993.1 | - | 2 | 589 | |||
BC032123.2 | - | 2 | 648 | 4853202 | ||
BC053665.1 | - | 3 | 400 | 6153096 | ||
BG720745.1 | - | 3 | 471 | |||
BQ050575.1 | - | 3 | 527 | |||
BX421356.2 | - | 4 | 716 | |||
CN430374.1 | - | 3 | 343 | |||
CN430377.1 | - | 3 | 681 |