Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135919.1 | NP_001129391.1 | 3 | 845 | ||
NM_181785.3 | NP_861450.1 | 3 | 845 | |||
XM_005266361.2 | XP_005266418.1 | 3 | 523 | |||
GenBank mRNA | AF315594.1 | - | 3 | 429 | ||
AK093932.1 | - | 2 | 1273 | |||
AK124604.1 | - | 1 | 1537 | |||
BC036662.1 | - | 3 | 807 | 4827143 | ||
BC060850.1 | - | 3 | 666 | 4820254 | ||
BC068556.1 | - | 3 | 673 | 4820792 | ||
BX537565.1 | - | 3 | 656 |