Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174976.2 | NP_777636.2 | 2 | 539 | ||
XM_006720119.3 | XP_006720182.1 | 2 | 769 | |||
XM_011536661.2 | XP_011534963.1 | 2 | 4408 | |||
GenBank mRNA | AK095612.1 | - | 2 | 539 | ||
AK294840.1 | - | 2 | 538 | |||
BC117676.2 | - | 1 | 364 | 40069732 | ||
DA496626.1 | - | 2 | 539 |