Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184739.1 | NP_001171668.1 | 4 | 1181 | ||
NM_001308067.1 | NP_001294996.1 | 4 | 876 | |||
NM_174944.3 | NP_777604.2 | 4 | 1151 | |||
XM_011536663.2 | XP_011534965.1 | 4 | 1033 | |||
XM_017021226.1 | XP_016876715.1 | 4 | 1419 | |||
GenBank mRNA | AF542390.1 | - | 2 | 1144 | ||
AK131042.1 | - | 4 | 1181 | |||
AY461663.1 | - | 4 | 1115 | |||
BC111088.1 | - | 4 | 813 | 40036837 | ||
BI825382.1 | - | 2 | 223 |