Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184739.1 | NP_001171668.1 | 1 | 389 | ||
NM_174944.3 | NP_777604.2 | 1 | 389 | |||
XM_017021226.1 | XP_016876715.1 | 1 | 585 | |||
GenBank mRNA | AK131042.1 | - | 1 | 389 | ||
AY461663.1 | - | 1 | 353 |