Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042367.1 | NP_001035826.1 | 3, 4 | 355 | ||
XM_017022083.1 | XP_016877572.1 | 2, 3 | 147 | |||
GenBank mRNA | BC055412.1 | - | 3, 4 | 336 | ||
BX113903.1 | - | 1, 2 | 66 | |||
DB465854.1 | - | 3, 4 | 355 |