Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000130.4 | NP_000121.2 | 11 | 1862 | ||
XM_017000660.1 | XP_016856149.1 | 10 | 1871 | |||
GenBank mRNA | AJ297254.1 | - | 1 | 106 | ||
AK226079.1 | - | 11 | 1780 | |||
AK291613.1 | - | 11 | 1812 | |||
M14335.1 | - | 11 | 1793 | |||
M16967.1 | - | 11 | 1807 |