Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143838.2 | NP_001137310.1 | 11 | 1543 | ||
NM_001284509.1 | NP_001271438.1 | 12 | 1630 | |||
NM_001284510.1 | NP_001271439.1 | 11 | 1552 | |||
NM_177550.4 | NP_808218.1 | 12 | 1681 | |||
GenBank mRNA | AK124317.1 | - | 1 | 3036 | ||
AK127797.1 | - | 12 | 1578 | |||
AK172779.1 | - | 5 | 534 | |||
AK172785.1 | - | 12 | 1626 | |||
AK297612.1 | - | 11 | 1497 | |||
AY151833.1 | - | 12 | 1605 | |||
BC104795.1 | - | 12 | 1637 | 8143798 | ||
BC112151.1 | - | 12 | 1637 | 8327619 | ||
BC143689.1 | - | 11 | 1499 | 9052203 | ||
BX647354.1 | - | 1 | 3786 | |||
CR749584.1 | - | 7 | 795 |