Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012232.5 | NP_036364.2 | 1 | 539 | ||
XM_005257242.3 | XP_005257299.1 | 1 | 539 | |||
GenBank mRNA | AF312393.1 | - | 1 | 736 | ||
AK000715.1 | - | 1 | 25 | |||
AK092406.1 | - | 2 | 340 | |||
AK314389.1 | - | 1 | 475 | |||
BC066123.1 | - | 1 | 475 | 5269172 | ||
BC073759.1 | - | 66 | 6572478 |