Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198476.3 | NP_940878.3 | 6 | 1929 | ||
XM_005258776.4 | XP_005258833.1 | 6 | 2499 | |||
XM_005258778.4 | XP_005258835.1 | 6 | 1918 | |||
XM_011526774.2 | XP_011525076.1 | 8 | 1976 | |||
XM_011526776.2 | XP_011525078.1 | 8 | 1904 | |||
XM_011526777.2 | XP_011525079.1 | 7 | 1859 | |||
XM_011526778.2 | XP_011525080.1 | 7 | 1787 | |||
XM_011526783.2 | XP_011525085.1 | 6 | 1586 | |||
XM_011526784.2 | XP_011525086.1 | 9 | 1299 | |||
XM_011526785.2 | XP_011525087.1 | 8 | 1385 | |||
XM_017026623.1 | XP_016882112.1 | 5 | 1469 | |||
XM_017026626.1 | XP_016882115.1 | 7 | 1832 | |||
XM_017026627.1 | XP_016882116.1 | 5 | 2385 | |||
GenBank mRNA | BC090894.1 | - | 6 | 1697 | 30377779 |