Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198478.3 | NP_940880.3 | 4 | 1471 | ||
XM_011526799.2 | XP_011525101.1 | 5 | 3710 | |||
XM_011526802.2 | XP_011525104.1 | 5 | 3449 | |||
XM_011526804.2 | XP_011525106.1 | 4 | 3272 | |||
XM_011526805.2 | XP_011525107.1 | 3 | 1328 | |||
GenBank mRNA | BC117228.1 | - | 1 | 887 | 40125779 | |
BC143792.1 | - | 1 | 887 | 9052306 |