Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145402.1 | NP_001138874.1 | 9 | 2029 | ||
GenBank mRNA | AK127979.1 | - | 7 | 2034 | ||
AL834316.1 | - | 1 | 811 |