Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178527.3 | NP_848622.2 | 13 | 1967 | ||
XM_011509426.1 | XP_011507728.1 | 13 | 1967 | |||
XM_017001065.1 | XP_016856554.1 | 13 | 1662 | |||
XM_017001066.1 | XP_016856555.1 | 13 | 1656 | |||
XM_017001067.1 | XP_016856556.1 | 13 | 1967 | |||
XM_017001068.1 | XP_016856557.1 | 12 | 1866 | |||
XM_017001069.1 | XP_016856558.1 | 12 | 1561 | |||
XM_017001070.1 | XP_016856559.1 | 12 | 1555 | |||
XM_017001071.1 | XP_016856560.1 | 13 | 1574 | |||
XM_017001072.1 | XP_016856561.1 | 11 | 1700 | |||
XM_017001073.1 | XP_016856562.1 | 12 | 1473 | |||
XM_017001074.1 | XP_016856563.1 | 11 | 1402 | |||
XM_017001075.1 | XP_016856564.1 | 6 | 729 | |||
XM_017001076.1 | XP_016856565.1 | 3 | 336 | |||
GenBank mRNA | AK128104.1 | - | 13 | 1967 | ||
AK302731.1 | - | 12 | 1683 | |||
BC042592.1 | - | 13 | 1914 | 4838901 | ||
BX648119.1 | - | 6 | 878 |