Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320996.1 | NP_001307925.1 | 1 | 323 | ||
NM_004102.4 | NP_004093.1 | 1 | 323 | |||
XM_011541007.2 | XP_011539309.1 | 1 | 653 | |||
GenBank mRNA | AK314122.1 | - | 1 | 49 | ||
BC007021.1 | - | 1 | 111 | 3934295 | ||
BF576953.1 | - | 1 | 112 | |||
BI752278.1 | - | 1 | 100 | |||
BP194697.1 | - | 1 | 323 | |||
BT006727.1 | - | 1 | 49 | |||
CN283506.1 | - | 1 | 102 | |||
CR456867.1 | - | 1 | 49 | |||
X56549.1 | - | 1 | 94 | |||
Y10255.1 | - | 1 | 53 |