Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SH2D6 SIRNA Assays ›
Gene Symbol:
SH2D6
Gene Name:
SH2 domain containing 6
Gene Aliases:
-
Chromosome Location:
Chr.2: 85418931 - 85437029 on Build GRCh38
UniGene ID:
Hs.209542
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s49776
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SH2D6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_201594.2
NP_963888.2
21
1951
XM_011532783.1
XP_011531085.1
14
1418
XM_011532796.2
XP_011531098.1
10
839
XM_017003834.1
XP_016859323.1
17
5299
XM_017003835.1
XP_016859324.1
17
4726
XM_017003836.1
XP_016859325.1
16
5050
XM_017003837.1
XP_016859326.1
17
5168
XM_017003838.1
XP_016859327.1
16
4787
XM_017003839.1
XP_016859328.1
16
2342
XM_017003840.1
XP_016859329.1
16
5398
XM_017003842.1
XP_016859331.1
18
5147
XM_017003843.1
XP_016859332.1
17
5032
XM_017003844.1
XP_016859333.1
18
4358
XM_017003845.1
XP_016859334.1
18
4800
XM_017003846.1
XP_016859335.1
18
5093
XM_017003847.1
XP_016859336.1
15
5318
XM_017003848.1
XP_016859337.1
15
5316
XM_017003849.1
XP_016859338.1
17
5078
XM_017003850.1
XP_016859339.1
17
4978
XM_017003851.1
XP_016859340.1
15
5339
XM_017003852.1
XP_016859341.1
14
1003
XM_017003858.1
XP_016859347.1
21
1925
XM_017003859.1
XP_016859348.1
14
1139
XM_017003860.1
XP_016859349.1
11
881
GenBank mRNA
AF450483.1
-
5
868
AK093312.1
-
21
1951
AY517502.1
-
21
1951
DB507279.3
-
7
673
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
immune response
transmembrane receptor protein tyrosine kinase signaling pathway
intracellular signal transduction
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0