Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001444.2 | NP_001435.1 | 2 | 253 | ||
GenBank mRNA | AK226036.1 | - | 2 | 202 | ||
AK311856.1 | - | 2 | 200 | |||
BC019385.2 | - | 2 | 186 | 3862100 | ||
BC070303.1 | - | 1 | 200 | 6496304 | ||
BT007449.1 | - | 1 | 140 | |||
CN278427.1 | - | 2 | 373 | |||
M94856.1 | - | 2 | 188 |