Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032504.1 | NP_115893.1 | 4 | 579 | ||
NM_182587.3 | NP_872393.3 | 4 | 579 | |||
XM_005246476.1 | XP_005246533.1 | 4 | 579 | |||
XM_011511004.1 | XP_011509306.1 | 4 | 579 | |||
XM_011511005.1 | XP_011509307.1 | 4 | 579 | |||
XM_011511006.1 | XP_011509308.1 | 4 | 579 | |||
XM_011511007.1 | XP_011509309.1 | 4 | 579 | |||
XM_011511008.1 | XP_011509310.1 | 4 | 579 | |||
XM_011511010.2 | XP_011509312.1 | 4 | 579 | |||
XM_011511012.1 | XP_011509314.1 | 4 | 579 | |||
XM_017003884.1 | XP_016859373.1 | 4 | 579 | |||
XM_017003885.1 | XP_016859374.1 | 4 | 579 | |||
XM_017003886.1 | XP_016859375.1 | 4 | 579 | |||
XM_017003887.1 | XP_016859376.1 | 4 | 579 | |||
XM_017003888.1 | XP_016859377.1 | 4 | 579 | |||
XM_017003889.1 | XP_016859378.1 | 4 | 579 | |||
XM_017003890.1 | XP_016859379.1 | 4 | 579 | |||
XM_017003891.1 | XP_016859380.1 | 3 | 598 | |||
XM_017003893.1 | XP_016859382.1 | 4 | 579 | |||
GenBank mRNA | AK090815.1 | - | 5 | 579 | ||
BC136690.1 | - | 4 | 550 | |||
BC136693.1 | - | 4 | 550 |