Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000136.2 | NP_000127.2 | 4, 5 | 615 | ||
NM_001243743.1 | NP_001230672.1 | 4, 5 | 557 | |||
NM_001243744.1 | NP_001230673.1 | 4, 5 | 615 | |||
XM_006717001.2 | XP_006717064.1 | 4, 5 | 597 | |||
XM_006717002.3 | XP_006717065.1 | 4, 5 | 597 | |||
XM_006717004.3 | XP_006717067.1 | 4, 5 | 597 | |||
XM_011518365.2 | XP_011516667.1 | 4, 5 | 590 | |||
XM_011518366.2 | XP_011516668.1 | 4, 5 | 597 | |||
GenBank mRNA | AK222871.1 | - | 4, 5 | 577 | ||
AK312548.1 | - | 4, 5 | 481 | |||
AU132608.1 | - | 4, 5 | 615 | |||
BC015748.1 | - | 4, 5 | 605 | 4864916 | ||
KU178051.1 | - | 3, 4 | 353 | |||
S64600.1 | - | 15 | ||||
X66893.1 | - | 4, 5 | 608 | |||
X66894.1 | - | 4, 5 | 526 |