Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308229.1 | NP_001295158.1 | 3 | 332 | ||
NM_173657.2 | NP_775928.1 | 4 | 459 | |||
XM_005247379.4 | XP_005247436.1 | 3 | 1304 | |||
XM_011512710.2 | XP_011511012.1 | 2 | 257 | |||
XM_011512711.2 | XP_011511013.1 | 3 | 436 | |||
GenBank mRNA | AF115515.1 | - | 3 | 295 | ||
AK055701.1 | - | 4 | 438 | |||
AK289890.1 | - | 3 | 332 | |||
AL832165.1 | - | 3 | 317 | |||
BC042198.1 | - | 3 | 294 | |||
BC064898.1 | - | 3 | 304 | 6008321 | ||
DA275323.1 | - | 3 | 332 |