Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207352.3 | NP_997235.3 | 10, 11 | 1706 | ||
XM_005262935.3 | XP_005262992.1 | 10, 11 | 1488 | |||
XM_017008037.1 | XP_016863526.1 | 9, 10 | 1464 | |||
GenBank mRNA | AK122600.1 | - | 10, 11 | 1441 | ||
AK126473.1 | - | 11, 12 | 1375 | |||
AK311390.1 | - | 4, 5 | 492 | |||
AY422002.1 | - | 10, 11 | 1706 | |||
BC041839.1 | - | 3, 4 | 1140 | |||
BC060857.1 | - | 10, 11 | 1572 | 30333559 | ||
BX648537.1 | - | 4, 5 | 637 | |||
BX648730.1 | - | 5, 6 | 5460 |