Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012761.2 | NP_001012779.2 | 4 | 1043 | ||
XM_005271966.3 | XP_005272023.1 | 4 | 899 | |||
XM_011543345.2 | XP_011541647.1 | 4 | 804 | |||
XM_011543346.2 | XP_011541648.1 | 2 | 1181 | |||
XM_011543347.2 | XP_011541649.2 | 3 | 727 | |||
XM_017009390.1 | XP_016864879.1 | 4 | 1291 | |||
XM_017009391.1 | XP_016864880.1 | 4 | 1189 | |||
GenBank mRNA | AK074887.1 | - | 4 | 1290 | ||
BC067736.1 | - | 4 | 1043 | 30340500 |