Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170692.1 | NP_001164163.1 | 5, 6 | 2282 | ||
NM_001170693.1 | NP_001164164.1 | 5, 6 | 2282 | |||
NM_205864.2 | NP_995586.1 | 4, 5 | 2064 | |||
GenBank mRNA | AF414185.1 | - | 4, 5 | 2063 | ||
AK097760.1 | - | 5, 6 | 1981 | |||
BC026188.1 | - | 5, 6 | 1955 | |||
BC026194.2 | - | 5, 6 | 1967 | 4826784 | ||
BC033545.1 | - | 5, 6 | 1967 | 4826548 |