Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181723.2 | NP_859074.1 | 9 | 982 | ||
XM_005273477.2 | XP_005273534.1 | 8 | 941 | |||
XM_006716333.3 | XP_006716396.1 | 9 | 999 | |||
XM_011544507.2 | XP_011542809.1 | 9 | 962 | |||
XM_011544508.2 | XP_011542810.1 | 10 | 1041 | |||
XM_017013336.1 | XP_016868825.1 | 8 | 923 | |||
XM_017013337.1 | XP_016868826.1 | 9 | 980 | |||
XM_017013338.1 | XP_016868827.1 | 10 | 1337 | |||
XM_017013339.1 | XP_016868828.1 | 10 | 1334 | |||
XM_017013340.1 | XP_016868829.1 | 8 | 927 | |||
XM_017013341.1 | XP_016868830.1 | 9 | 1295 | |||
XM_017013342.1 | XP_016868831.1 | 7 | 794 | |||
GenBank mRNA | BC032868.1 | - | 9 | 937 | ||
BC045553.1 | - | 9 | 981 | 5262603 |