Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144877.2 | NP_001138349.1 | 18 | 1899 | ||
NM_173690.4 | NP_775961.2 | 19 | 1968 | |||
GenBank mRNA | AF086497.1 | - | 5 | 513 | ||
AK093983.1 | - | 18 | 1858 | |||
AK098024.1 | - | 19 | 1905 | |||
BC104030.2 | - | 19 | 1896 | 40025626 | ||
BC104031.1 | - | 18 | 1827 | 40025627 | ||
BC104032.1 | - | 18 | 1745 | 40025629 |