Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 3 | 689 | ||
NR_120531.1 | 3 | 689 | ||||
XM_005250297.3 | XP_005250354.1 | 3 | 684 | |||
XM_011516114.1 | XP_011514416.1 | 2 | 348 | |||
XM_017012093.1 | XP_016867582.1 | 2 | 348 | |||
GenBank mRNA | AK223406.1 | - | 3 | 588 | ||
AK300223.1 | - | 3 | 478 | |||
AK312752.1 | - | 3 | 605 | |||
BC014142.2 | - | 3 | 559 | 4662011 | ||
CD014094.1 | - | 3 | 571 | |||
M15182.1 | - | 3 | 584 |