Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 1846 | ||
GenBank mRNA | AY168647.2 | - | 1 | 1428 | ||
BC017510.1 | - | 1 | 1221 | |||
BC028316.1 | - | 1 | 945 | |||
BC036931.1 | - | 1 | 924 | |||
BC046919.1 | - | 1 | 1287 | |||
BC110824.1 | - | 1 | 1428 | 5169320 |