Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022725.3 | NP_073562.1 | 1 | 488 | ||
GenBank mRNA | AF181994.1 | - | 1 | 461 | ||
AF181995.1 | - | 1 | 457 | |||
AK001716.2 | - | 1 | 470 | |||
AK023153.1 | - | 1 | 470 | |||
AK223277.1 | - | 1 | 462 | |||
BC035346.1 | - | 1 | 482 | |||
BC047028.1 | - | 1 | 460 | 5538578 | ||
BC063038.1 | - | 1 | 488 | 5239725 | ||
BC093867.1 | - | 1 | 478 | 7939712 | ||
BC101807.1 | - | 1 | 478 | 8069313 | ||
BI522448.1 | - | 1 | 488 | |||
BX640996.1 | - | 1 | 480 |