Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134649.2 | NP_001128121.1 | 7 | 708 | ||
NM_001134650.1 | NP_001128122.1 | 7 | 584 | |||
NM_001134651.1 | NP_001128123.1 | 6 | 575 | |||
NM_001282886.1 | NP_001269815.1 | 7 | 701 | |||
NM_173359.4 | NP_775495.1 | 7 | 669 | |||
XM_011533651.2 | XP_011531953.1 | 4 | 428 | |||
XM_017006278.1 | XP_016861767.1 | 8 | 715 | |||
XM_017006279.1 | XP_016861768.1 | 8 | 702 | |||
XM_017006280.1 | XP_016861769.1 | 8 | 849 | |||
XM_017006281.1 | XP_016861770.1 | 6 | 577 | |||
XM_017006282.1 | XP_016861771.1 | 8 | 657 | |||
GenBank mRNA | AK126999.1 | - | 6 | 457 | ||
AK313656.1 | - | 7 | 669 | |||
BC031289.2 | - | 5 | 392 | 5299499 | ||
BC068443.1 | - | 7 | 701 | 4820931 | ||
BX648727.1 | - | 3 | 189 | |||
DA227887.1 | - | 7 | 559 | |||
DR159502.1 | - | 6 | 574 |