Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134649.2 | NP_001128121.1 | 6, 7 | 624 | ||
NM_001134650.1 | NP_001128122.1 | 6, 7 | 500 | |||
NM_001134651.1 | NP_001128123.1 | 5, 6 | 491 | |||
NM_001282886.1 | NP_001269815.1 | 6, 7 | 617 | |||
NM_173359.4 | NP_775495.1 | 6, 7 | 585 | |||
XM_011533651.2 | XP_011531953.1 | 3, 4 | 344 | |||
XM_017006278.1 | XP_016861767.1 | 7, 8 | 631 | |||
XM_017006279.1 | XP_016861768.1 | 7, 8 | 618 | |||
XM_017006280.1 | XP_016861769.1 | 7, 8 | 765 | |||
XM_017006281.1 | XP_016861770.1 | 5, 6 | 493 | |||
XM_017006282.1 | XP_016861771.1 | 7, 8 | 573 | |||
GenBank mRNA | AK126999.1 | - | 5, 6 | 373 | ||
AK313656.1 | - | 6, 7 | 585 | |||
BC031289.2 | - | 4, 5 | 308 | 5299499 | ||
BC068443.1 | - | 6, 7 | 617 | 4820931 | ||
BX648727.1 | - | 2, 3 | 105 | |||
DA227887.1 | - | 6, 7 | 475 | |||
DA394405.1 | - | 6, 7 | 500 | |||
DR159502.1 | - | 5, 6 | 490 |