Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000202.7 | NP_000193.1 | 6 | 1029 | ||
NM_001166550.3 | NP_001160022.1 | 6 | 985 | |||
NM_006123.4 | NP_006114.1 | 6 | 1029 | |||
NR_104128.1 | 6 | 1029 | ||||
GenBank mRNA | AK294541.1 | - | 6 | 940 | ||
BC006170.1 | - | 6 | 887 | 4025980 | ||
BX647357.1 | - | 6 | 883 | |||
L40586.1 | - | 6 | 939 | |||
M58342.1 | - | 6 | 936 |