Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSC2 SIRNA Assays ›
Gene Symbol:
TSC2
Gene Name:
tuberous sclerosis 2
Gene Aliases:
LAM, PPP1R160, TSC4
Chromosome Location:
Chr.16: 2047804 - 2088720 on Build GRCh38
UniGene ID:
Hs.90303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s502595
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000548.4
NP_000539.2
5
604
NM_001077183.2
NP_001070651.1
5
604
NM_001114382.2
NP_001107854.1
5
604
NM_001318827.1
NP_001305756.1
4
493
NM_001318829.1
NP_001305758.1
4
437
NM_001318832.1
NP_001305761.1
5
505
XM_005255529.4
XP_005255586.2
5
491
XM_005255531.4
XP_005255588.2
5
491
XM_011522636.2
XP_011520938.1
5
491
XM_011522637.2
XP_011520939.1
5
491
XM_011522638.2
XP_011520940.2
4
584
XM_011522639.2
XP_011520941.1
5
491
XM_011522640.2
XP_011520942.1
5
491
XM_017023615.1
XP_016879104.1
5
491
XM_017023616.1
XP_016879105.1
5
491
XM_017023617.1
XP_016879106.1
4
584
XM_017023618.1
XP_016879107.1
5
491
GenBank mRNA
AB210000.1
-
5
510
AK295672.1
-
4
324
AK295728.1
-
5
505
AK299343.1
-
4
374
BC150300.1
-
5
479
8860381
BF194994.1
-
2
111
KJ534954.1
-
4
404
KJ534955.1
-
4
404
KJ534958.1
-
4
404
KJ534982.1
-
4
404
KJ534984.1
-
4
404
KJ535011.1
-
4
404
KJ535038.1
-
4
1209
KJ535039.1
-
4
404
KJ535091.1
-
4
404
X75621.1
-
4
422
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
negative regulation of protein kinase activity
protein import into nucleus
endocytosis
cell cycle arrest
heart development
protein localization
negative regulation of cell proliferation
negative regulation of phosphatidylinositol 3-kinase signaling
viral process
vesicle-mediated transport
regulation of endocytosis
negative regulation of Wnt signaling pathway
negative regulation of TOR signaling
protein kinase B signaling
positive regulation of GTPase activity
regulation of insulin receptor signaling pathway
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
positive chemotaxis
regulation of small GTPase mediated signal transduction
regulation of cell cycle
negative regulation of protein kinase B signaling
GTPase activator activity
protein binding
phosphatase binding
small GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0