Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006638.2 | NP_001006639.1 | 9 | 676 | ||
NM_001163989.1 | NP_001157461.1 | 9 | 825 | |||
NM_001163990.1 | NP_001157462.1 | 8 | 565 | |||
NM_175738.4 | NP_783865.1 | 9 | 1069 | |||
XM_005257288.4 | XP_005257345.1 | 9 | 694 | |||
GenBank mRNA | AK054846.1 | - | 6 | 1804 | ||
AK057069.1 | - | 3 | 220 | |||
AK098068.1 | - | 9 | 676 | |||
AK127073.1 | - | 8 | 1829 | |||
AK290202.1 | - | 9 | 1045 | |||
AK296062.1 | - | 9 | 664 | |||
AK296172.1 | - | 8 | 528 | |||
AK303442.1 | - | 9 | 728 | |||
BC016615.1 | - | 9 | 622 | 4520191 | ||
BC040547.1 | - | 9 | 1055 | 5287038 |