Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021175.3 | NP_066998.1 | 3 | 521 | ||
GenBank mRNA | AF131292.1 | - | 3 | 335 | ||
AF147355.1 | - | 1 | 41 | |||
AF309489.1 | - | 3 | 360 | |||
AJ277280.1 | - | 3 | 327 | |||
AK096110.1 | - | 1 | 2565 | |||
AY358669.1 | - | 3 | 329 | |||
BC020612.1 | - | 3 | 339 | 4715540 | ||
BP306149.1 | - | 3 | 523 |