Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005167.5 | NP_005158.5 | 5 | 909 | ||
GenBank mRNA | AB451327.1 | - | 4 | 264 | ||
AB451471.1 | - | 4 | 264 | |||
AK093270.1 | - | 5 | 889 | |||
AL133644.1 | - | 6 | 1673 | |||
BC011803.2 | - | 5 | 833 | |||
BC064628.1 | - | 5 | 839 | |||
BC073828.1 | - | 5 | 901 | 5244363 |