Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012302.2 | NP_001012302.2 | 20 | 1987 | ||
XM_011520053.2 | XP_011518355.1 | 20 | 2083 | |||
GenBank mRNA | AK026923.1 | - | 20 | 1934 | ||
AK074088.1 | - | 13 | 4604 | |||
AK096874.1 | - | 11 | 1275 | |||
AL832289.1 | - | 9 | 1909 | |||
BC047946.1 | - | 1 | 588 | |||
BC128557.1 | - | 20 | 1955 | 40122929 | ||
BC144415.1 | - | 19 | 2013 |