Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008223.1 | NP_001008224.1 | 2 | 1809 | ||
XM_011538270.2 | XP_011536572.1 | 3 | 1614 | |||
GenBank mRNA | AF466366.1 | - | 2 | 1809 | ||
BC110812.1 | - | 2 | 1807 | 3913171 |