Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029861.2 | NP_001025032.1 | 1 | 944 | ||
GenBank mRNA | BC042883.1 | - | 1 | 946 | 5270224 |