Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178558.4 | NP_848653.2 | 4 | 1526 | ||
XM_006715959.2 | XP_006716022.1 | 5 | 1802 | |||
XM_017012126.1 | XP_016867615.1 | 7 | 2108 | |||
XM_017012127.1 | XP_016867616.1 | 6 | 1920 | |||
XM_017012128.1 | XP_016867617.1 | 6 | 1990 | |||
XM_017012129.1 | XP_016867618.1 | 5 | 1652 | |||
GenBank mRNA | AK074911.1 | - | 1 | 806 | ||
AK122937.1 | - | 4 | 1487 | |||
BC030700.1 | - | 4 | 1510 | 4837196 |