Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282863.1 | NP_001269792.1 | 3 | 611 | ||
NM_001317942.1 | NP_001304871.1 | 3 | 330 | |||
NM_178565.4 | NP_848660.3 | 4 | 1009 | |||
XM_011517018.1 | XP_011515320.1 | 4 | 1025 | |||
XM_011517019.1 | XP_011515321.1 | 4 | 1025 | |||
XM_017013395.1 | XP_016868884.1 | 4 | 998 | |||
GenBank mRNA | AK123023.1 | - | 4 | 972 | ||
AK123027.1 | - | 2 | 139 | |||
AY358166.1 | - | 4 | 662 | |||
BC027938.1 | - | 3 | 330 | 5201681 | ||
BC036554.1 | - | 3 | 612 | 5265957 |