Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178566.4 | NP_848661.1 | 5 | 687 | ||
XM_017014682.1 | XP_016870171.1 | 3 | 471 | |||
XM_017014683.1 | XP_016870172.1 | 6 | 727 | |||
XM_017014684.1 | XP_016870173.1 | 6 | 659 | |||
XM_017014685.1 | XP_016870174.1 | 6 | 607 | |||
XM_017014686.1 | XP_016870175.1 | 7 | 788 | |||
XM_017014687.1 | XP_016870176.1 | 6 | 735 | |||
XM_017014688.1 | XP_016870177.1 | 6 | 817 | |||
XM_017014689.1 | XP_016870178.1 | 6 | 793 | |||
XM_017014690.1 | XP_016870179.1 | 7 | 677 | |||
XM_017014691.1 | XP_016870180.1 | 6 | 3202 | |||
XM_017014692.1 | XP_016870181.1 | 6 | 467 | |||
XM_017014693.1 | XP_016870182.1 | 7 | 524 | |||
GenBank mRNA | AK292960.1 | - | 5 | 494 | ||
BC042558.1 | - | 5 | 681 | 4837003 |