Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012980.2 | NP_001012998.2 | 3 | 1755 | ||
XM_011530940.2 | XP_011529242.1 | 4 | 2189 | |||
GenBank mRNA | BC043215.1 | - | 3 | 1230 | 5295273 | |
BC126401.1 | - | 3 | 1215 | 8992117 |